Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromoindol-3-propionic acid 97%

Supplier: AMBEED, INC
Warning

A397168-100MG A397168-1G A397168-250MG
77101-844EA 56 USD InStock
77101-844 77101-846 77101-848
5-Bromoindol-3-propionic acid 97%
5-Bromoindol-3-propionic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₀BrNO₂
MDL Number: MFCD02325723
CAS Number: 54904-23-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty