Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3-chloroanisole 98%

Supplier: AMBEED, INC
Warning

A397379-250MG A397379-25G A397379-5G
77101-850EA 7 USD InStock
77101-850 77101-852 77101-854
2-Bromo-3-chloroanisole 98%
2-Bromo-3-chloroanisole
Precaución: for Research Use Only


Formula: C₇H₆BrClO
MW: 221.479 g/mol
MDL Number: MFCD18390300
CAS Number: 174913-08-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty