Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Bromomethyl)-1-methyl-1H-indazole 95%

Supplier: AMBEED, INC
Danger

A413358-100MG A413358-250MG
77102-048EA 277 USD InStock
77102-048 77102-050
3-(Bromomethyl)-1-methyl-1H-indazole 95%
3-(Bromomethyl)-1-methyl-1H-indazole
Precaución: for Research Use Only


Formula: C₉H₉BrN₂
MDL Number: MFCD22412488
CAS Number: 174180-57-5
UN: 1759
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty