Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-fluoropyridine 98%

Supplier: AMBEED, INC
Warning

A416634-100G A416634-10G A416634-25G A416634-500G A416634-5G
77102-086EA 58 USD InStock
77102-086 77102-088 77102-090 77102-092 77102-094
2-Chloro-6-fluoropyridine 98%
2-Chloro-6-fluoropyridine
Precaución: for Research Use Only


Formula: C₅H₃ClFN
MW: 131.54
MDL Number: MFCD00077483
CAS Number: 20885-12-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty