Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1-{[(tert-Butoxycarbonyl)amino]methyl}cyclohexyl)acetic acid 97%

Supplier: AMBEED, INC
Warning

A421442-1G A421442-250MG A421442-5G
77102-136EA 42 USD InStock
77102-136 77102-138 77102-140
(1-{[(tert-Butoxycarbonyl)amino]methyl}cyclohexyl)acetic acid 97%
(1-{[(tert-Butoxycarbonyl)amino]methyl}cyclohexyl)acetic acid
Precaución: for Research Use Only


Formula: C₁₄H₂₅NO₄
MW: 271.36 g/mol
MDL Number: MFCD03839829
CAS Number: 227626-60-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty