Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-(pyrrolidin-1-yl)pyrimidine 97%

Supplier: AMBEED, INC
Warning

A423150-100MG A423150-1G A423150-250MG A423150-5G
77102-152EA 8 USD InStock
77102-152 77102-154 77102-156 77102-158
4-Chloro-2-(pyrrolidin-1-yl)pyrimidine 97%
4-Chloro-2-(pyrrolidin-1-yl)pyrimidine
Precaución: for Research Use Only


Formula: C₈H₁₀ClN₃
MDL Number: MFCD09607721
CAS Number: 33852-01-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty