Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-([1,1'-Biphenyl]-4-yl)-4-chloro-6-phenyl-1,3,5-triazine 98%

Supplier: AMBEED, INC
Warning

A427588-10G A427588-1G A427588-25G A427588-5G
77102-214EA 60 USD InStock
77102-214 77102-216 77102-218 77102-220
2-([1,1'-Biphenyl]-4-yl)-4-chloro-6-phenyl-1,3,5-triazine 98%
2-([1,1'-Biphenyl]-4-yl)-4-chloro-6-phenyl-1,3,5-triazine
Precaución: for Research Use Only


Formula: C₂₁H₁₄ClN₃
MDL Number: MFCD27979856
CAS Number: 1472062-94-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty