Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoropyridine 98%

Supplier: AMBEED, INC
Danger

A435189-1G A435189-250MG A435189-25G A435189-5G A435189-100G A435189-10G
77102-358EA 9 USD InStock
77102-358 77102-360 77102-362 77102-364 77686-183 77686-184
2,3-Difluoropyridine 98%
2,3-Difluoropyridine
Precaución: for Research Use Only


Formula: C₅H₃F₂N
MDL Number: MFCD04114143
CAS Number: 1513-66-2
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty