Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Bromo-p-toluonitrile 98%

Supplier: AMBEED, INC
Danger

A439623-10G A439623-25G A439623-5G
77102-408EA 7 USD InStock
77102-408 77102-410 77102-412
α-Bromo-p-toluonitrile 98%
α-Bromo-p-toluonitrile
Precaución: for Research Use Only


Formula: C₈H₆BrN
MW: 196.05 g/mol
Melting Pt: 115
MDL Number: MFCD00001829
CAS Number: 17201-43-3
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty