Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9,9-Bis(4-amino-3-chlorophenyl)fluorene 98%

Supplier: AMBEED, INC
Danger

A440537-10G A440537-1G A440537-5G
77102-432EA 60 USD InStock
77102-432 77102-434 77102-436
9,9-Bis(4-amino-3-chlorophenyl)fluorene 98%
9,9-Bis(4-amino-3-chlorophenyl)fluorene
Precaución: for Research Use Only


Formula: C₂₅H₁₈Cl₂N₂
MDL Number: MFCD10000948
CAS Number: 107934-68-9
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty