Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-((4-(2-Cyanophenyl)piperazin-1-yl)methyl)-3-methylbenzamide maleate 95%

Supplier: AMBEED, INC
Warning

A447591-100MG A447591-10MG A447591-1MG A447591-25MG A447591-50MG A447591-5MG
77102-484EA 123 USD InStock
77102-484 77102-486 77102-488 77102-490 77102-492 77102-494
N-((4-(2-Cyanophenyl)piperazin-1-yl)methyl)-3-methylbenzamide maleate 95%
N-((4-(2-Cyanophenyl)piperazin-1-yl)methyl)-3-methylbenzamide maleate
Precaución: for Research Use Only


Formula: C₂₄H₂₆N₄O₅
MDL Number: MFCD01321067
CAS Number: 630117-19-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty