Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Benzyloxy)-3-methylphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A449582-1G A449582-250MG A449582-5G
77102-512EA 107 USD InStock
77102-512 77102-514 77102-516
2-(Benzyloxy)-3-methylphenylboronic acid 98%
2-(Benzyloxy)-3-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₅BO₃
MW: 242.08 g/mol
MDL Number: MFCD06801732
CAS Number: 177190-68-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty