Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)pyridine 98%

Supplier: AMBEED, INC
Danger

A458022-10G A458022-1G A458022-25G A458022-5G
77102-566EA 18 USD InStock
77102-566 77102-568 77102-570 77102-572
2-(Trifluoromethyl)pyridine 98%
2-(Trifluoromethyl)pyridine
Precaución: for Research Use Only


Formula: C₆H₄F₃N
MDL Number: MFCD02183562
CAS Number: 368-48-9
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty