Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-4-(boc-amino)-L-phenylalanine 95%

Supplier: AMBEED, INC
Warning

A458632-100G A458632-1G A458632-250MG
77102-574EA 1839 USD InStock
77102-574 77102-576 77102-578
Fmoc-4-(boc-amino)-L-phenylalanine 95%
Fmoc-4-(boc-amino)-L-phenylalanine
Precaución: For research use only.


Formula: C₂₉H₃₀N₂O₆
MDL Number: MFCD00237007
CAS Number: 174132-31-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty