Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9,9-Dimethyl-9H-9-silafluorene 98%

Supplier: AMBEED, INC
Warning

A459074-100MG A459074-1G A459074-250MG A459074-5G
77102-580EA 20 USD InStock
77102-580 77102-582 77102-584 77102-586
9,9-Dimethyl-9H-9-silafluorene 98%
9,9-Dimethyl-9H-9-silafluorene
Precaución: for Research Use Only


Formula: C₁₄H₁₄Si
MW: 210.35 g/mol
MDL Number: MFCD00093801
CAS Number: 13688-68-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty