Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4,5,5-Tetramethyl-2-(2-methyl-4-(trifluoromethoxy)phenyl)-1,3,2-dioxaborolane 96%

Supplier: AMBEED, INC
Warning

A459607-100MG A459607-1G A459607-250MG A459607-5G
77102-610EA 57 USD InStock
77102-610 77102-612 77102-614 77102-616
4,4,5,5-Tetramethyl-2-(2-methyl-4-(trifluoromethoxy)phenyl)-1,3,2-dioxaborolane 96%
4,4,5,5-Tetramethyl-2-(2-methyl-4-(trifluoromethoxy)phenyl)-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₄H₁₈BF₃O₃
MW: 302.09 g/mol
MDL Number: MFCD11867879
CAS Number: 1309980-29-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty