Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-N-(tert-butyl)benzenesulfonamide 98%

Supplier: AMBEED, INC
Warning

A467738-10G A467738-1G A467738-250MG A467738-25G A467738-5G
77102-702EA 80 USD InStock
77102-702 77102-704 77102-706 77102-708 77102-710
4-Amino-N-(tert-butyl)benzenesulfonamide 98%
4-Amino-N-(tert-butyl)benzenesulfonamide
Precaución: for Research Use Only


Formula: C₁₀H₁₆N₂O₂S
MW: 228.32 g/mol
MDL Number: MFCD03002128
CAS Number: 209917-48-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty