Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-[3-Chloro-5-(trifluoromethyl)phenyl]propionic acid 95%

Supplier: AMBEED, INC
Warning

A471888-100MG A471888-1G A471888-250MG A471888-5G
77102-722EA 12 USD InStock
77102-722 77102-724 77102-726 77102-728
3-[3-Chloro-5-(trifluoromethyl)phenyl]propionic acid 95%
3-[3-Chloro-5-(trifluoromethyl)phenyl]propionic acid
Precaución: for Research Use Only


Formula: C₁₀H₈ClF₃O₂
MW: 252.618 g/mol
MDL Number: MFCD09025401
CAS Number: 916420-78-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty