Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-((2-Chloroethyl)amino)ethanol hydrochloride 95%

Supplier: AMBEED, INC
Warning

A502931-100MG A502931-1G A502931-250MG
77103-010EA 91 USD InStock
77103-010 77103-012 77103-014
2-((2-Chloroethyl)amino)ethanol hydrochloride 95%
2-((2-Chloroethyl)amino)ethanol hydrochloride
Precaución: for Research Use Only


Formula: C₄H₁₁Cl₂NO
MDL Number: MFCD28559188
CAS Number: 2576-29-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty