Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Bromo-3-chloro-4-fluorotoluene 98%

Supplier: AMBEED, INC
Danger

A503571-100G A503571-10G A503571-1G A503571-25G A503571-5G
77103-026EA 104 USD InStock
77103-026 77103-028 77103-030 77103-032 77103-034
α-Bromo-3-chloro-4-fluorotoluene 98%
α-Bromo-3-chloro-4-fluorotoluene
Precaución: for Research Use Only


Formula: C₇H₅BrClF
MW: 223.47
Boiling Pt: 223
Melting Pt: 37
MDL Number: MFCD01631551
CAS Number: 192702-01-5
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty