Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxyquinolin-6-ylboronic acid 95%

Supplier: AMBEED, INC
Warning

A514303-100MG A514303-1G A514303-250MG
77103-142EA 39 USD InStock
77103-142 77103-144 77103-146
2-Methoxyquinolin-6-ylboronic acid 95%
2-Methoxyquinolin-6-ylboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₀BNO₃
MDL Number: MFCD18384109
CAS Number: 1191061-58-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty