Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-chloro-1-fluorobenzene 98%

Supplier: AMBEED, INC
Warning

A517286-100G A517286-10G A517286-1G A517286-25G A517286-500G A517286-5G
77103-184EA 75 USD InStock
77103-184 77103-186 77103-188 77103-190 77103-192 77103-194
2-Bromo-4-chloro-1-fluorobenzene 98%
2-Bromo-4-chloro-1-fluorobenzene
Precaución: for Research Use Only


Formula: C₆H₃BrClF
MW: 209.44
MDL Number: MFCD00672934
CAS Number: 1996-30-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty