Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glyoxylic acid 50% in water

Supplier: AMBEED, INC
Warning

A521642-1000G A521642-100G A521642-25G A521642-500G
77103-216EA 18 USD InStock
77103-216 77103-218 77103-220 77103-222
Glyoxylic acid 50% in water
Glyoxylic acid
Precaución: for Research Use Only


Formula: C₂H₂O₃
MDL Number: MFCD00006958
CAS Number: 298-12-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty