Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-(isoquinolin-1-yl)acetate 95%

Supplier: AMBEED, INC
Warning

A522205-100MG A522205-1G A522205-250MG A522205-50MG
77103-224EA 41 USD InStock
77103-224 77103-226 77103-228 77103-230
Methyl 2-(isoquinolin-1-yl)acetate 95%
Methyl 2-(isoquinolin-1-yl)acetate
Precaución: for Research Use Only


Formula: C₁₂H₁₁NO₂
MDL Number: MFCD13194757
CAS Number: 69582-93-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty