Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dicyclohexylamine (R)-6-(((allyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)hexanoate 98%

Supplier: AMBEED, INC
Warning

A523414-1G A523414-250MG A523414-5G
77103-236EA 44 USD InStock
77103-236 77103-238 77103-240
Dicyclohexylamine (R)-6-(((allyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)hexanoate 98%
Dicyclohexylamine (R)-6-(((allyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)hexanoate
Precaución: for Research Use Only


Formula: C₂₇H₄₉N₃O₆
MDL Number: MFCD01862281
CAS Number: 327156-94-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty