Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glycine dimethylamide 98%

Supplier: AMBEED, INC
Danger

A526122-1G A526122-250MG A526122-25G
77103-270EA 68 USD InStock
77103-270 77103-272 77686-359
Glycine dimethylamide 98%
Glycine dimethylamide
Precaución: for Research Use Only


Formula: C₄H₁₀N₂O
MDL Number: MFCD06655265
CAS Number: 1857-19-8
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty