Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-((2,5-Difluorobenzyl)oxy)phenoxy)-5-ethoxyaniline 98%

Supplier: AMBEED, INC
Warning

A527418-1MG A527418-2MG A527418-5MG
77103-276EA 17 USD InStock
77103-276 77103-278 77103-280
2-(4-((2,5-Difluorobenzyl)oxy)phenoxy)-5-ethoxyaniline 98%
2-(4-((2,5-Difluorobenzyl)oxy)phenoxy)-5-ethoxyaniline
Precaución: for Research Use Only


Formula: C₂₁H₁₉F₂NO₃
MDL Number: MFCD07772188
CAS Number: 223104-29-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty