Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Tosylpiperidine 95+%

Supplier: AMBEED, INC
Warning

A533111-1G A533111-250MG A533111-5G
77103-354EA 167 USD InStock
77103-354 77103-356 77103-358
1-Tosylpiperidine 95+%
1-Tosylpiperidine
Precaución: for Research Use Only


Formula: C₁₂H₁₇NO₂S
MDL Number: MFCD00159367
CAS Number: 4703-22-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty