Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromomethyl-4-fluorobenzeneboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Danger

A534764-100MG A534764-1G A534764-250MG A534764-5G
77103-360EA 37 USD InStock
77103-360 77103-362 77103-364 77103-366
3-Bromomethyl-4-fluorobenzeneboronic acid pinacol ester 97%
3-Bromomethyl-4-fluorobenzeneboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₇BBrFO₂
MDL Number: MFCD23098825
CAS Number: 1544739-25-4
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty