Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-chlorothiophene-2-carboxylate 98%

Supplier: AMBEED, INC
Warning

A535272-100G A535272-10G A535272-1G A535272-25G A535272-5G
77103-368EA 148 USD InStock
77103-368 77103-370 77103-372 77103-374 77103-376
Methyl-3-chlorothiophene-2-carboxylate 98%
Methyl-3-chlorothiophene-2-carboxylate
Precaución: for Research Use Only


Formula: C₆H₅ClO₂S
MDL Number: MFCD00068135
CAS Number: 88105-17-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty