Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-fluoro-6-iodoaniline 98%

Supplier: AMBEED, INC
Warning

A536626-100G A536626-10G A536626-1G A536626-250MG A536626-25G A536626-5G
77103-384EA 227 USD InStock
77103-384 77103-386 77103-388 77103-390 77103-392 77103-394
4-Chloro-2-fluoro-6-iodoaniline 98%
4-Chloro-2-fluoro-6-iodoaniline
Precaución: for Research Use Only


Formula: C₆H₄ClFIN
MW: 271.46 g/mol
MDL Number: MFCD01318122
CAS Number: 216393-67-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty