Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-Phe(4-NH2)-OMe 95%

Supplier: AMBEED, INC
Warning

A539277-100MG A539277-25MG A539277-5G
77103-424EA 7 USD InStock
77103-424 77103-426 77569-052
Boc-Phe(4-NH2)-OMe 95%
Boc-Phe(4-NH2)-OMe
Precaución: For research use only.


Formula: C₁₅H₂₂N₂O₄
MW: 294.35 g/mol
MDL Number: MFCD11655908
CAS Number: 65615-90-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty