Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-bromo-5-chloroisonicotinate 95%

Supplier: AMBEED, INC
Danger

A566218-100MG A566218-1G A566218-5G
77103-656EA 9 USD InStock
77103-656 77103-658 77686-442
Methyl 2-bromo-5-chloroisonicotinate 95%
Methyl 2-bromo-5-chloroisonicotinate
Precaución: for Research Use Only


Formula: C₇H₅BrClNO₂
MW: 250.477 g/mol
MDL Number: MFCD14698101
CAS Number: 1214336-33-0
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty