Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Methoxymethyl)-pyrrolidine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A578198-1G A578198-250MG A578198-5G
77103-724EA 45 USD InStock
77103-724 77103-726 77103-728
3-(Methoxymethyl)-pyrrolidine hydrochloride 98%
3-(Methoxymethyl)-pyrrolidine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₃NO·ClH
MDL Number: MFCD09997928
CAS Number: 1185303-79-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty