Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentafluorophenyl trifluoromethanesulfonate 97%

Supplier: AMBEED, INC
Danger

A580660-100MG A580660-1G A580660-250MG A580660-5G
77103-758EA 7 USD InStock
77103-758 77103-760 77103-762 77103-764
Pentafluorophenyl trifluoromethanesulfonate 97%
Pentafluorophenyl trifluoromethanesulfonate
Precaución: for Research Use Only


Formula: C₇F₈O₃S
MDL Number: MFCD07784319
CAS Number: 60129-85-3
UN: 3082
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty