Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-2-Methyl-3-phenylacrylaldehyde 97%

Supplier: AMBEED, INC
Warning

A581012-1000G A581012-100G A581012-25G A581012-500G
77103-768EA 102 USD InStock
77103-768 77103-770 77103-772 77103-774
(E)-2-Methyl-3-phenylacrylaldehyde 97%
(E)-2-Methyl-3-phenylacrylaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₁₀O
MDL Number: MFCD00006976
CAS Number: 15174-47-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty