Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-8-methoxyquinoline 98%

Supplier: AMBEED, INC
Warning

A583077-100MG A583077-1G A583077-250MG
77103-806EA 8 USD InStock
77103-806 77103-808 77103-810
4-Hydroxy-8-methoxyquinoline 98%
4-Hydroxy-8-methoxyquinoline
Precaución: for Research Use Only


Formula: C₁₀H₉NO₂
MW: 175.184 g/mol
MDL Number: MFCD02684163
CAS Number: 21269-34-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty