Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-3-iodo-1H-pyrazolo[4,3-c]pyridine 95%

Supplier: AMBEED, INC
Warning

A588012-100MG A588012-250MG
77103-868EA 40 USD InStock
77103-868 77103-870
6-Bromo-3-iodo-1H-pyrazolo[4,3-c]pyridine 95%
6-Bromo-3-iodo-1H-pyrazolo[4,3-c]pyridine
Precaución: for Research Use Only


Formula: C₆H₃BrIN₃
MDL Number: MFCD22380432
CAS Number: 1357945-49-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty