Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Daidzin 98+%

Supplier: AMBEED, INC
Warning

A590254-100MG A590254-10MG A590254-1G A590254-1MG A590254-250MG
77103-888EA 64 USD InStock
77103-888 77103-890 77103-892 77103-894 77103-896
Daidzin 98+%
Daidzin
Precaución: for Research Use Only


Formula: C₂₁H₂₀O₉
MDL Number: MFCD00017466
CAS Number: 552-66-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty