Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1lambda(6),2,5-Thiadiazepane-1,1-dione 97%

Supplier: AMBEED, INC
Warning

A591259-100MG A591259-250MG
77103-908EA 275 USD InStock
77103-908 77686-501
1lambda(6),2,5-Thiadiazepane-1,1-dione 97%
1lambda(6),2,5-Thiadiazepane-1,1-dione
Precaución: for Research Use Only


Formula: C₄H₁₀N₂O₂S
MDL Number: MFCD20662817
CAS Number: 410545-38-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty