Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-3-chloro-2-fluorobenzeneboronic acid 95+%

Supplier: AMBEED, INC
Warning

A599208-100MG A599208-1G A599208-250MG
77104-016EA 5 USD InStock
77104-016 77104-018 77104-020
6-Bromo-3-chloro-2-fluorobenzeneboronic acid 95+%
6-Bromo-3-chloro-2-fluorobenzeneboronic acid
Precaución: for Research Use Only


Formula: C₆H₄BBrClFO₂
MW: 253.261 g/mol
MDL Number: MFCD11111906
CAS Number: 1451392-87-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty