Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-nitro-1H-indole-6-carboxylate 97+%

Supplier: AMBEED, INC
Warning

A604458-100MG A604458-1G A604458-250MG
77104-094EA 10 USD InStock
77104-094 77104-096 77104-098
Methyl 4-nitro-1H-indole-6-carboxylate 97+%
Methyl 4-nitro-1H-indole-6-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₈N₂O₄
MDL Number: MFCD09880080
CAS Number: 1000343-62-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty