Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 2,4,5-trifluorobenzoate 95%

Supplier: AMBEED, INC
Warning

A620895-100G A620895-25G A620895-5G
77104-222EA 64 USD InStock
77104-222 77104-224 77104-226
Sodium 2,4,5-trifluorobenzoate 95%
Sodium 2,4,5-trifluorobenzoate
Precaución: for Research Use Only


Formula: C₇H₂F₃NaO₂
MDL Number: MFCD03094399
CAS Number: 522651-48-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty