Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Biphenylyl)-4,6-dichloro-1,3,5-triazine 98%

Supplier: AMBEED, INC
Warning

A623854-1G A623854-25G A623854-5G
77104-248EA 8 USD InStock
77104-248 77104-250 77104-252
2-(4-Biphenylyl)-4,6-dichloro-1,3,5-triazine 98%
2-(4-Biphenylyl)-4,6-dichloro-1,3,5-triazine
Precaución: for Research Use Only


Formula: C₁₅H₉Cl₂N₃
MDL Number: MFCD28556865
CAS Number: 10202-45-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty