Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-Methoxy-5-(methylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%

Supplier: AMBEED, INC
Warning

A656123-100MG A656123-1G A656123-250MG
77104-538EA 39 USD InStock
77104-538 77104-540 77104-542
2-(3-Methoxy-5-(methylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%
2-(3-Methoxy-5-(methylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₄H₂₁BO₃S
MDL Number: MFCD16618974
CAS Number: 1256360-25-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty