Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-bromo-5-hydroxybenzoate 95%

Supplier: AMBEED, INC
Warning

A658496-100MG A658496-1G A658496-250MG A658496-5G
77104-556EA 7 USD InStock
77104-556 77104-558 77104-560 77104-562
Methyl 2-bromo-5-hydroxybenzoate 95%
Methyl 2-bromo-5-hydroxybenzoate
Precaución: for Research Use Only


Formula: C₈H₇BrO₃
MW: 231.043 g/mol
MDL Number: MFCD10566797
CAS Number: 154607-00-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty