Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2,3,6-trifluorobenzene 98%

Supplier: AMBEED, INC
Warning

A664322-100G A664322-1G A664322-25G A664322-5G
77104-600EA 374 USD InStock
77104-600 77104-602 77104-604 77104-606
1-Bromo-2,3,6-trifluorobenzene 98%
1-Bromo-2,3,6-trifluorobenzene
Precaución: for Research Use Only


Formula: C₆H₂BrF₃
MDL Number: MFCD00236214
CAS Number: 176793-04-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty