Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 2-cyclohexylhydrazinecarboxylate 95+%

Supplier: AMBEED, INC
Warning

A664974-100MG A664974-1G A664974-250MG A664974-5G
77104-630EA 7 USD InStock
77104-630 77104-632 77104-634 77104-636
tert-Butyl 2-cyclohexylhydrazinecarboxylate 95+%
tert-Butyl 2-cyclohexylhydrazinecarboxylate
Precaución: for Research Use Only


Formula: C₁₁H₂₂N₂O₂
MW: 214.30 g/mol
MDL Number: MFCD11044977
CAS Number: 60295-21-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty