Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoro-3-methylphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A665657-100MG A665657-1G A665657-250MG A665657-5G
77104-642EA 37 USD InStock
77104-642 77104-644 77104-646 77104-648
2,6-Difluoro-3-methylphenylboronic acid 98%
2,6-Difluoro-3-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₇BF₂O₂
MW: 171.937 g/mol
MDL Number: MFCD09265479
CAS Number: 1586045-40-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty