Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9,10-Bis[N,N-di(p-tolyl)amino]anthracene 98%

Supplier: AMBEED, INC
Warning

A674242-100MG A674242-250MG
77104-742EA 33 USD InStock
77104-742 77104-744
9,10-Bis[N,N-di(p-tolyl)amino]anthracene 98%
9,10-Bis[N,N-di(p-tolyl)amino]anthracene
Precaución: for Research Use Only


Formula: C₄₂H₃₆N₂
MDL Number: MFCD12022465
CAS Number: 177799-16-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty